Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Butyne-1-ol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: But-2-yn-1-ol2-Butyn-1-ol1-Hydroxy-2-butyne

OR14505-25G OR14505-100G OR14505-5G
77404-984EA 13 USD InStock
77404-984 77404-982 77404-980
2-Butyne-1-ol 97%
2-Butyne-1-ol


Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142…143 °C
Melting Pt: –2 °C
Density: 0.938 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty