Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Morpholine 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR13977-100G
77404-776EA 13 USD InStock
77404-776
Morpholine 99%
Morpholine


Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 129 °C
Melting Pt: –7 to –5 °C
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8+3,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty