Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3'-(Trifluoromethyl)acetophenone 96%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-[3-(Trifluoromethyl)phenyl]ethan-1-onem-(Trifluoromethyl)acetophenone

PC49336-500G PC49336-100G
77430-430EA 82 USD InStock
77430-430 77430-432
3'-(Trifluoromethyl)acetophenone 96%
3'-(Trifluoromethyl)acetophenone


Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198…200 °C
Density: 1.235 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty