Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Lactic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (R)-(-)-Lactic acid(R)-(-)-2-Hydroxypropanoic acidD-Lactic Acid(R)-(-)-2-Hydroxypropionic acid(R)-2-Hydroxypropanoic acid

OR960721-50G OR960721-100G OR960721-500G
77425-840EA 149 USD InStock
77425-840 77425-838 77425-836
D-(-)-Lactic acid
D(-)-Lactic acid


Formula: C₃H₆O₃
MW: 90.08 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00068313
CAS Number: 10326-41-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty