Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluorophenyl isocyanate 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,4-Difluorophenylisocyanate

PC2875H-100G PC2875H-5G PC2875H-25G
77413-950EA 204 USD InStock
77413-950 77413-952 77413-954
2,4-Difluorophenyl isocyanate 98%
2,4-Difluorophenyl isocyanate
  • May form small amount of sediment.


Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 42 °C
Density: 1.309 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001997
CAS Number: 59025-55-7
UN: 2478
ADR: 3+6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty