Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7734-100G PC7734-25G PC7734-500G
77418-156EA 686 USD InStock
77418-156 77418-152 77418-154
2-Fluorobenzylamine 95%
2-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 73…75 °C/13 mmHg
Density: 1.099 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty