Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,5,6-Tetrafluorobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (2,3,5,6-Tetrafluorophenyl)methanol

PC1300-25G PC1300-1G PC1300-5G
77426-938EA 22 USD InStock
77426-938 77426-940 77426-936
2,3,5,6-Tetrafluorobenzyl alcohol 98%
2,3,5,6-Tetrafluorobenzyl alcohol


Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 220…225 °C
Melting Pt: 34…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00792428
CAS Number: 4084-38-2
EINECS: 443-840-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty