Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-o-toluidine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Fluoro-2-methylaniline2-Amino-4-fluorotoluene

PC3822-25G PC3822-500G PC3822-100G
77414-928EA 25 USD InStock
77414-928 77414-930 77414-932
5-Fluoro-o-toluidine 98%
5-Fluoro-o-toluidine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 98…100 °C/15 mmHg
Melting Pt: 37…40 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty