Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Fluorobenzyl Alcohol(2-Fluorophenyl)methanol

PC3640-100G PC3640-25G
77428-898EA 44 USD InStock
77428-898 77428-900
2-Fluorobenzyl alcohol 97%
2-Fluorobenzyl alcohol


Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 97…101 °C/21 mmHg
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004601
CAS Number: 446-51-5
EINECS: 207-170-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty