Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-fluorobenzoic acid

Supplier: APOLLO SCIENTIFIC

PC3918-5G
77415-012EA 14 USD InStock
77415-012
2-Bromo-6-fluorobenzoic acid
2-Bromo-6-fluorobenzoic acid


Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD01569539
CAS Number: 2252-37-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty