Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isonicotinic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Pyridinecarboxylic acidPyridine-4-carboxylic acid4-Picolinic acid

OR23064-500G OR23064-25G
77407-056EA 13 USD InStock
77407-056 77407-058
Isonicotinic acid 99%
Isonicotinic acid


Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 260 °C/15 mmHg
Melting Pt: 310…315 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty