Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) fluoride 99,9%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Ytterbium trifluorideYtterbium fluoride

PC8022-10G
77418-338EA 49 USD InStock
77418-338
Ytterbium(III) fluoride 99,9%
Ytterbium (III) fluoride


Formula: F₃Yb
MW: 230.04 g/mol
Boiling Pt: 2200 °C
Melting Pt: 1157 °C
Density: 8.168 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049615
CAS Number: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty