Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(IV) oxide 99.9+%

Supplier: APOLLO SCIENTIFIC
Synonyms: Tin dioxideStannic OxideTin(iv)oxide

IN3673-500G IN3673-100G
77401-584EA 108 USD InStock
77401-584 77401-582
Tin(IV) oxide 99.9+%
Tin (IV) oxide


Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: 1800…900 °C
Melting Pt: 1630 °C
Density: 6.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty