Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

Supplier: APOLLO SCIENTIFIC
Synonyms: trans-1,2-Cyclohexanediaminetetraacetic acid monohydratetrans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrateCyDTA monohydratetrans-Cyclohex-1,2-ylenediaminetetra(acetic acid) monohydratetrans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydratetrans-(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, MonohydrateCDTA monohydrateTitriplex® IV(1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate

OR926084-25G OR926084-100G
77425-176EA 68 USD InStock
77425-176 77425-178
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate


Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty