Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Chloro-4'-fluoroacetanilide 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: N-(2-Chloro-4-fluorophenyl)acetamide

PC1814-50G PC1814-250G
77427-276EA 13 USD InStock
77427-276 77427-274
2'-Chloro-4'-fluoroacetanilide 99%
2'-Chloro-4'-fluoroacetanilide


Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Melting Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00042594
CAS Number: 399-35-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty