Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Paracetamol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: APAPParacetamolumAcetaminophenN-(4-Hydroxyphenyl)acetamide4-AcetamidophenolN-Acetyl-4-aminophenol4'-Hydroxyacetanilide

OR3962-5KG OR3962-250G OR3962-1KG
77420-804EA 502 USD InStock
77420-804 77420-800 77420-802
Paracetamol 98%
Paracetamol


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 168…172 °C
Density: 1.293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty