Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω,ω-Difluoroacetophenone 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,2-Difluoro-1-phenylethanone2,2-Difluoroacetophenoneα,α-Difluoroacetophenone

PC6686-5G PC6686-1G
77417-492EA 206 USD InStock
77417-492 77417-490
ω,ω-Difluoroacetophenone 95%
ω,ω-Difluoroacetophenone


Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 63…65 °C/11 mmHg
Melting Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD06246879
CAS Number: 395-01-7
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty