Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Tolylacetic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (3-Methylphenyl)acetic acid2-(m-Tolyl)acetic acid

OR4364-250G OR4364-50G OR4364-10G
77421-484EA 74 USD InStock
77421-484 77421-488 77421-486
m-Tolylacetic acid 98%
m-Tolylacetic acid


Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004340
CAS Number: 621-36-3
EINECS: 210-683-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty