Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Chloromethyl)trimethylsilane 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR3470-1G
77409-272EA 13 USD InStock
77409-272
(Chloromethyl)trimethylsilane 98%
(Chloromethyl)trimethylsilane


Formula: ClH₂CSi(CH₃)₃
MW: 122.67 g/mol
Boiling Pt: 98…99 °C
Density: 0.879 g/cm³ (25 °C)
Flash Pt: –4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5
UN: 2924
ADR: 3+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty