Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Trifluoromethyl)trimethylsilane 0,5 M in THF 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: trimethyl(trifluoromethyl)silane

PC3953-5ML PC3953-25ML PC3953-100ML
77415-036EA 13 USD InStock
77415-036 77415-034 77415-032
(Trifluoromethyl)trimethylsilane 0,5 M in THF 98%
(Trifluoromethyl)trimethylsilane


Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 40 °C
Density: 0.895 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty