Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Bis(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: α,α,α,α',α',α'-Hexafluoro-3,5-xylenol

PC3112-100G PC3112-25G
77428-532EA 34 USD InStock
77428-532 77428-530
3,5-Bis(trifluoromethyl)phenol 98%
3,5-Bis(trifluoromethyl)phenol


Formula: C₈H₄F₆O
MW: 230.11 g/mol
Boiling Pt: 97…98 °C/50 mmHg
Melting Pt: 20…21 °C
Density: 1.511 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty