Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitro-1H-indazole 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-NINA5-Nitro-1H-indazole5-Nitroindazole

OR9104-100G OR9104-25G
77424-862EA 61 USD InStock
77424-862 77424-864
5-Nitro-1H-indazole 95%
5-Nitro-1H-indazole


Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00005693
CAS Number: 5401-94-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty