Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromopicolinic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Bromopyridine-2-carboxylic acid3-Bromo-2-pyridinecarboxylic acid

OR8877-1G
77424-652EA 13 USD InStock
77424-652
3-Bromopicolinic acid
3-Bromopicolinic acid


Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 141…144 °C
Density: 1.813 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01320380
CAS Number: 30683-23-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty