Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Curcumin 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

OR24598-5G OR24598-100G OR24598-25G
77407-506EA 42 USD InStock
77407-506 77407-508 77407-504
Curcumin 97%
Curcumin


Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Freezer
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty