Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluoroacetone trihydrate 95%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Perfluoroacetone trihydrate

PC4571-100G PC4571-25G
77415-628EA 115 USD InStock
77415-628 77415-630
Hexafluoroacetone trihydrate 95%
Hexafluoroacetone trihydrate


Formula: C₃F₆O·3H₂O
MW: 220.07 g/mol
Boiling Pt: 105…108 °C
Melting Pt: 18…21 °C
Density: 1.579 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149061
CAS Number: 34202-69-2
EINECS: 211-676-3
UN: 2552
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty