Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Nitrophenyl)ethanol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Nitrophenethyl alcoholp-Nitrophenethyl Alcohol

OR1836-25G OR1836-100G
77406-284EA 67 USD InStock
77406-284 77406-282
2-(4-Nitrophenyl)ethanol 98%
2-(4-Nitrophenyl)ethanol


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 177 °C/16 mmHg
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00010202
CAS Number: 100-27-6
EINECS: 202-835-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty