Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: PTAD4-Phenyl-1,2,4-triazoline-3,5-dione4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione

OR939123-1G OR939123-5G
77412-058EA 60 USD InStock
77412-058 77412-060
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione 95%
4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione


Formula: C₈H₅N₃O₂
MW: 175.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003148
CAS Number: 4233-33-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty