Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-amino-4-methylthiazole-5-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 2-amino-4-methylthiazole-5-carboxylate

OR9899-25G
77425-996EA 36 USD InStock
77425-996
Ethyl-2-amino-4-methylthiazole-5-carboxylate 98%
Ethyl 2-amino-4-methylthiazole-5-carboxylate


Formula: C₇H₁₀N₂O₂S
MW: 186.22 g/mol
Melting Pt: 172…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00123414
CAS Number: 7210-76-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty