Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloro-4-fluorobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2453-25G PC2453-1G PC2453-5G
77427-648EA 47 USD InStock
77427-648 77427-652 77427-650
3,5-Dichloro-4-fluorobenzoic acid 98%
3,5-Dichloro-4-fluorobenzoic acid


Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840463
CAS Number: 98191-30-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty