Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydropyran-2-methanol 98%

Supplier: APOLLO SCIENTIFIC
Synonyms: 2-(Hydroxymethyl)tetrahydro-2H-pyran

OR5628-25G OR5628-5G
77423-158EA 161 USD InStock
77423-158 77423-160
Tetrahydropyran-2-methanol 98%
Tetrahydropyran-2-methanol


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 186…189 °C
Density: 1.027 g/cm³ (25 °C)
Flash Pt: >76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006624
CAS Number: 100-72-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty