Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylthio)phenol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-(Methylmercapto)phenol

OR0778-100G OR0778-50G OR0778-500G
77402-228EA 178 USD InStock
77402-228 77402-232 77402-230
4-(Methylthio)phenol 98%
4-(Methylthio)phenol


Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 153…156 °C/20 mmHg
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002351
CAS Number: 1073-72-9
EINECS: 214-031-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty