Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Boc-3-aminopyrrolidine 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 3-Aminopyrrolidine, N1-BOC protected1-Boc-3-Aminopyrrolidine

OR6139-25G OR6139-1G OR6139-5G
77410-742EA 26 USD InStock
77410-742 77410-740 77410-738
(±)-1-Boc-3-aminopyrrolidine 97%
(±)-1-Boc-3-aminopyrrolidine


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Density: 1.022 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861220
CAS Number: 186550-13-0
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty