Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-1H-indazole 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Iodoindazole

OR305076-1G OR305076-5G
77417-260EA 13 USD InStock
77417-260 77417-650
3-Iodo-1H-indazole 98%
3-Iodo-1H-indazole


Formula: C₇H₅IN₂
MW: 244.0325 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Ambient
MDL Number: MFCD04972469
CAS Number: 66607-27-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty