Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-piperidine-4-carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acidN-BOC-4-Piperidinecarboxylic acidBoc-Inp-OH1-Boc-isonipecotic acid

OR5410-100G OR5410-500G OR5410-25G
77422-846EA 51 USD InStock
77422-846 77422-848 77422-850
1-Boc-piperidine-4-carboxylic acid 98%
1-Boc-piperidine-4-carboxylic acid


Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00076999
CAS Number: 84358-13-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty