Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Difluoromethoxy)iodobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(Difluoromethoxy)-4-iodobenzene

PC2847R-25G PC2847R-100G PC2847R-5G
77427-924EA 65 USD InStock
77427-924 77427-922 77427-926
4-(Difluoromethoxy)iodobenzene 98%
4-(Difluoromethoxy)iodobenzene


Formula: C₇H₅F₂IO
MW: 270.0153 g/mol
Boiling Pt: 217…219 °C
Density: 1.871 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00085008
CAS Number: 128140-82-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty