Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) trifluoromethanesulfonate

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Nickel(II) trifluoromethanesulfonateTrifluoromethanesulphonic acid Nickel(II) SaltTrifluoromethanesulfonic acid Nickel(II) SaltNickel triflateNickel(II) trifluoromethanesulphonateNickel(II) Triflate

PC3387-25G PC3387-5G PC3387-1G
77428-810EA 13 USD InStock
77428-810 77428-808 77428-806
Nickel(II) trifluoromethanesulfonate
Nickel(II) trifluoromethanesulfonate


Formula: C₂F₆NiO₆S₂
MW: 356.82 g/mol
Melting Pt: 100…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00673740
CAS Number: 60871-84-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty