Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-3-(trimethylsilyl)-1-propanesulfonate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Sodium 2,2-Dimethyl-2-silapentane-5-sulphonate3-(Trimethylsilyl)-1-propanesulfonic acid Sodium SaltSodium 2,2-Dimethyl-2-silapentane-5-sulfonateDSS3-(Trimethylsilyl)-1-propanesulphonic acid Sodium Salt

DE985-1G
77401-344EA 58 USD InStock
77401-344
Sodium-3-(trimethylsilyl)-1-propanesulfonate
Sodium-3-(trimethylsilyl)-1-propanesulfonate
Not Deuterated.


Formula: C₆H₁₆O₃SSi
MW: 218.32 g/mol
Melting Pt: 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007537
CAS Number: 2039-96-5
EINECS: 218-031-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty