Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%

Supplier: APOLLO SCIENTIFIC

PC3861-10G PC3861-5G PC3861-1G
77414-980EA 83 USD InStock
77414-980 77414-982 77414-978
2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%
2,3-Dibromo-5-(Trifluoromethyl)Pyridine


Formula: C₆H₂Br₂F₃N
MW: 304.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687233
CAS Number: 79623-38-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty