Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Benzylhydroxylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR15742-100G OR15742-25G
77405-546EA 141 USD InStock
77405-546 77405-548
O-Benzylhydroxylamine 97%
O-Benzylhydroxylamine


Formula: C₇H₉NO
Boiling Pt: 118…119 °C/30 mmHg
Density: 1.058 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00221709
CAS Number: 622-33-3
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty