Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-5-methylindole-2-carboxylate 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 5-methylindole-2-carboxylate

OR322120-10G OR322120-5G OR322120-1G
77420-056EA 120 USD InStock
77420-056 77420-060 77420-058
Ethyl-5-methylindole-2-carboxylate 97%
Ethyl-5-methylindole-2-carboxylate


Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022703
CAS Number: 16382-15-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty