Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-methoxyaniline

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Amino-4-fluoroanisole6-Fluoro-m-anisidine

PC49529-25G PC49529-1G PC49529-5G
77430-592EA 34 USD InStock
77430-592 77430-594 77430-596
2-Fluoro-5-methoxyaniline
2-Fluoro-5-methoxyaniline


Formula: C₇H₈FNO
MW: 141.143 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226283
CAS Number: 62257-15-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty