Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-methoxy-5-nitrobenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC502390-1G PC502390-250MG
77430-844EA 456 USD InStock
77430-844 77430-846
3-Fluoro-4-methoxy-5-nitrobenzoic acid 95%
3-Fluoro-4-methoxy-5-nitrobenzoic acid


Formula: C₈H₆FNO₅
MW: 215.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12173014
CAS Number: 577-39-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty