Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Difluoro-4-hydrazinopyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,6-Difluoro-4-hydrazinylpyridine

PC9683-1G PC9683-100MG PC9683-250MG
77418-722EA 20 USD InStock
77418-722 77418-724 77418-726
2,6-Difluoro-4-hydrazinopyridine 97%
2,6-Difluoro-4-hydrazinopyridine


Formula: C₅H₅F₂N₃
MW: 145.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436095
CAS Number: 837364-94-0
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty