Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Genie® Multi-Microplate, 230 V

Supplier: Electron Microscopy Sciences
61042-AU 61042-EU 61042-BR 61042-SW 61042-20
102093-402EA 1616.56 USD InStock
102093-402 102093-404 102093-406 102093-408 102093-410
Genie® Multi-Microplate, 230 V
Shakers and Mixers Microplate Shakers
Microplate Mixer

  • 230 V model
  • Aggressive mixing speed from 750 to 3200 rpm for complete and uniform mixing in any microplate format
  • May be used to shake up to four microplates, an optional accessory tray is available to double capacity and shake up to eight microplates at one time
  • Single capacity microplate mixer is also available
  • Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity
  • Speed and orbit combine to offer true vortexing action in each well of a microplate
  • Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats
  • Built-in timer operates from 1 to 60 minutes or continuous for hands-free use
  • May be used in cold rooms and incubators for temperature sensitive applications

Additional accessories for the Multi-Microplate Genie® available are listed with Microplate Genie®.

Caution: 230 V model. Corresponding models with voltage suitable for use in US (102093-398 or 102093-400) are also available, please see separate product page.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty