Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-4-nitrobenzotrifluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Chloro-4-nitrobenzotrifluoride1-Chloro-2-nitro-5-(trifluoromethyl)benzene2-Chloro-1-nitro-4-(trifluoromethyl)benzene

PC8117-25G PC8117-1G
77433-560EA 18 USD InStock
77433-560 77433-562
3-Chloro-4-nitrobenzotrifluoride 97%
3-Chloro-4-nitrobenzotrifluoride


Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 54 °C/ 0.4 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00272564
CAS Number: 402-11-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty