Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2,4,6-trifluoropyrimidine 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC9980-1G
77434-692EA 13 USD InStock
77434-692
5-Chloro-2,4,6-trifluoropyrimidine 95%
5-Chloro-2,4,6-trifluoropyrimidine


Formula: C₄ClF₃N₂
Boiling Pt: 116 °C
Density: 1.626 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00191933
CAS Number: 697-83-6
EINECS: 211-807-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty