Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-5-nitroanisole 97%

Supplier: APOLLO SCIENTIFIC

PC902146-10G PC902146-1G
77433-862EA 604 USD InStock
77433-862 77433-864
3-Fluoro-5-nitroanisole 97%
3-Fluoro-5-nitroanisole


Formula: C₇H₆FNO₃
MW: 171.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848484
CAS Number: 7087-60-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty