Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Alanine

Supplier: VWR
Synonyms: L-α-AlanineL(+)-AlanineAα-AlanineAlanineH-Ala-OH(S)-(+)-AlanineH-L-Ala-OH

VWRVBRGT106-5KG
80760-316EA 244.84 USD InStock
80760-316
L-Alanine
L-Alanine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00064410
CAS Number: 56-41-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty