Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Compact Shakers, 220 V

Supplier: Yamato
MK-161-220V MK-201D-220V
76550-268EA 3236.52 USD InStock
76550-268 76550-272
Compact Shakers, 220 V
Shakers and Mixers Rotating/Rocking Shakers
The compact design of this shaker saves precious space.

  • 220 V model
  • Rotary shaking and reciprocate range: 30 mm
  • Various shaking modes included
  • Shaking frequency: 20 to ~200 rpm, random adjustment
  • Digital display of shaking frequency
  • Equipped with shaking timer

It features changeable rotary and reciprocate motions to mix, extract, and stir samples.

A shaking timer is provided, and a digital display indicates shaking frequency. The unit works with IN602CSW incubator for shaking incubation.

Accessories are easy to add and remove.

Caution: 220 V model. Corresponding models with voltage suitable for use in US (76550-266 or 76550-270) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty