Would you like to log in again?
Ward's® High Speed Microcentrifuges, 230 V
Supplier: MOLECULAR ARTS CORP SE
Ward's®
C2417-E_WARD
470320-562EA 2493 USD InStock
470320-562
Ward's® High Speed Microcentrifuges, 230 V
Centrifuges Microcentrifuges
Touch screen, high speed centrifugation.
  • 230 V model
  • Full-color touch screen
  • One-button program recall
  • Combination rotor accepts tubes and strips
  • Compact design
  • Two-year warranty
Ward's High speed centrifuge features a next-generation control panel with touch screen. It keeps samples cool, even during extended runs at the maximum speed setting by a proprietary air flow system that moves air evenly and quietly throughout the chamber.
The unique combination-rotor accepts both standard (1.5 or 2.0 ml) microtubes and PCR tubes/strips, making this the perfect partner for any molecular biology laboratory.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (470320-564) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty